Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ihh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010544.2 | NP_034674.1 | 2 | 999 | ||
GenBank mRNA | AK090147.1 | - | 2 | 794 | ||
AK144322.1 | - | 2 | 1022 | |||
BC046984.1 | - | 2 | 999 | 6477118 | ||
U85610.1 | - | 2 | 859 | |||
X76291.1 | - | 2 | 337 |