Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008406.3 | NP_032432.2 | 20 | 2445 | ||
GenBank mRNA | AK149500.1 | - | 20 | 2446 | ||
BC013465.1 | - | 20 | 2429 | |||
BC028814.1 | - | 20 | 2445 | 5099284 | ||
X70391.1 | - | 20 | 2425 |