Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010593.2 | NP_034723.1 | 2, 3 | 369 | ||
GenBank mRNA | AK145895.1 | - | 2, 3 | 354 | ||
AK170934.1 | - | 2, 3 | 342 | |||
AK171501.1 | - | 343 | ||||
BC040757.1 | - | 2, 3 | 318 | 4216772 | ||
BC094461.1 | - | 2, 3 | 322 | 4459882 | ||
CD555608.1 | - | 2, 3 | 369 |