Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081133.2 | NP_001074602.1 | 19, 20 | 3265 | ||
GenBank mRNA | AB610968.1 | - | 20, 21 | 3353 | ||
AK129401.1 | - | 7, 8 | 1469 | |||
AK142909.1 | - | 1, 2 | 565 | |||
AK160835.1 | - | 3, 4 | 1485 |