Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290995.1 | NP_001277924.1 | 12 | 1344 | ||
NM_008441.2 | NP_032467.2 | 11 | 1291 | |||
NM_207682.2 | NP_997565.2 | 11 | 1291 | |||
GenBank mRNA | AB023656.1 | - | 11 | 1175 | ||
AF090190.1 | - | 11 | 970 | |||
AF131865.1 | - | 12 | 1262 | |||
AK134946.1 | - | 11 | 1327 | |||
D17577.1 | - | 11 | 1244 |