Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008474.2 | NP_032500.2 | 1 | 118 | ||
GenBank mRNA | AY028607.1 | - | 1 | 75 | ||
BC114971.1 | - | 1 | 71 | 40039508 | ||
BY779643.1 | - | 1 | 118 |