Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008522.3 | NP_032548.2 | 16 | 2062 | ||
GenBank mRNA | AK036491.1 | - | 16 | 2063 | ||
AK144556.1 | - | 16 | 2063 | |||
AK150799.1 | - | 16 | 2063 | |||
AK151822.1 | - | 16 | 2063 | |||
AK164781.1 | - | 16 | 2064 | |||
BC006904.1 | - | 16 | 2070 | 3600033 | ||
BC008530.1 | - | 16 | 2043 | |||
BC009662.1 | - | 12 | 1493 | |||
CT010339.1 | - | 16 | 2028 | |||
D88510.1 | - | 16 | 2062 | |||
FJ538998.1 | - | 17 | 2028 | |||
J03298.1 | - | 16 | 2031 |