Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 8 | 798 | ||
GenBank mRNA | AK144883.1 | - | 8 | 775 | ||
AK154285.1 | - | 8 | 752 | |||
AK160205.1 | - | 8 | 764 | |||
AK163150.1 | - | 8 | 711 | |||
AK167345.1 | - | 8 | 763 | |||
BC001984.1 | - | 8 | 734 | 3489973 | ||
BC004673.1 | - | 2 | 218 | |||
BC005802.1 | - | 7 | 807 | 3707919 | ||
U50734.1 | - | 8 | 716 |