Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010772.1 | NP_034902.1 | 4 | 1262 | ||
GenBank mRNA | AK209702.1 | - | 2 | 113 | ||
BC004758.1 | - | 6 | 799 | |||
DQ150586.1 | - | 3 | 976 | |||
L04649.1 | - | 4 | 1235 |