Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008565.3 | NP_032591.3 | 10 | 1520 | ||
GenBank mRNA | AK011743.1 | - | 10 | 1455 | ||
AK036653.1 | - | 10 | 1480 | |||
AK088796.1 | - | 10 | 1440 | |||
AK089999.1 | - | 10 | 1424 | |||
AK135422.1 | - | 10 | 1422 | |||
AK141132.1 | - | 10 | 1487 | |||
AK141644.1 | - | 10 | 1521 | |||
AK146662.1 | - | 10 | 1443 | |||
AK151499.1 | - | 10 | 1434 | |||
AK157725.1 | - | 10 | 1422 | |||
AK220442.1 | - | 10 | 1422 | |||
BC013094.1 | - | 10 | 1476 | 3584640 | ||
D26089.1 | - | 10 | 1426 |