Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008585.2 | NP_032611.2 | 1 | 81 | ||
GenBank mRNA | AK093914.1 | - | 1 | 42 | ||
AK143906.1 | - | 1 | 38 | |||
AK164734.1 | - | 1 | 36 | |||
BC015258.1 | - | 1 | 81 | 4241252 | ||
M74897.1 | - | 1 | 80 |