Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mitf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113198.1 | NP_001106669.1 | 4 | 750 | ||
NM_001178049.1 | NP_001171520.1 | 4 | 637 | |||
NM_008601.3 | NP_032627.1 | 3 | 441 | |||
GenBank mRNA | AB221657.1 | - | 3 | 303 | ||
AK052895.1 | - | 4 | 609 | |||
AK154189.1 | - | 4 | 751 | |||
AK155350.1 | - | 4 | 751 | |||
BC108976.2 | - | 3 | 405 | 40047440 | ||
Z23066.1 | - | 3 | 432 |