Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008617.2 | NP_032643.2 | 9 | 1005 | ||
GenBank mRNA | AK002305.1 | - | 9 | 1022 | ||
AK135162.1 | - | 9 | 1018 | |||
AK160553.1 | - | 9 | 988 | |||
AK167809.1 | - | 9 | 988 | |||
BC023482.1 | - | 9 | 1005 | 5251192 | ||
DQ402950.1 | - | 8 | 855 | |||
M16229.1 | - | 9 | 964 |