Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161817.2 | NP_001155289.1 | 16 | 1710 | ||
NM_001290982.1 | NP_001277911.1 | 16 | 1710 | |||
NM_010863.4 | NP_034993.2 | 16 | 1710 | |||
GenBank mRNA | AK141207.1 | - | 16 | 1685 | ||
AK149409.1 | - | 16 | 1657 | |||
AK161062.1 | - | 16 | 1674 | |||
AK169513.1 | - | 6 | 660 | |||
BC026826.1 | - | 16 | 1658 | |||
BC046300.1 | - | 16 | 1656 | 6487332 | ||
BC054786.1 | - | 16 | 1660 | 6821232 | ||
L00923.1 | - | 16 | 1682 | |||
X69987.1 | - | 16 | 1630 |