Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008681.2 | NP_032707.2 | 13, 14 | 999 | ||
GenBank mRNA | AA795988.1 | - | 5, 6 | 292 | ||
AK004548.1 | - | 13, 14 | 999 | |||
AK098075.1 | - | 13, 14 | 998 | |||
AK161860.1 | - | 13, 14 | 1001 | |||
AK170281.1 | - | 13, 14 | 1000 | |||
AK170439.1 | - | 13, 14 | 998 | |||
AK170636.1 | - | 13, 14 | 998 | |||
AK170809.1 | - | 13, 14 | 999 | |||
BC015282.1 | - | 13, 14 | 954 | 4237943 | ||
BC071235.1 | - | 13, 14 | 1007 | 4242075 | ||
BC141246.1 | - | 13, 14 | 890 | 9055858 | ||
BC171968.1 | - | 14, 15 | 890 | 9054254 | ||
U52073.1 | - | 13, 14 | 972 | |||
U60593.1 | - | 13, 14 | 997 |