Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008682.2 | NP_032708.2 | 14 | 1940 | ||
GenBank mRNA | AK010591.1 | - | 14 | 1928 | ||
AK048393.1 | - | 14 | 1892 | |||
AK153746.1 | - | 14 | 1942 | |||
BC052430.1 | - | 14 | 1909 | 6836583 | ||
BC066870.1 | - | 14 | 1877 | 30476383 | ||
D10712.1 | - | 14 | 1882 |