Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081981.2 | NP_001075450.1 | 2 | 1494 | ||
NM_001081982.2 | NP_001075451.1 | 2 | 819 | |||
NM_001297601.1 | NP_001284530.1 | 2 | 1494 | |||
NM_010906.3 | NP_035036.1 | 2 | 819 | |||
GenBank mRNA | AK158568.1 | - | 2 | 1497 | ||
BC003766.1 | - | 2 | 541 | 3491917 | ||
BC090626.1 | - | 2 | 521 | |||
S81451.1 | - | 1 | 505 | |||
U57636.1 | - | 1 | 496 | |||
Y07688.1 | - | 2 | 821 | |||
Y07689.1 | - | 2 | 623 |