Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010927.3 | NP_035057.1 | 9 | 1112 | ||
GenBank mRNA | AF065919.1 | - | 9 | 1109 | ||
AF065920.1 | - | 9 | 1109 | |||
AF065921.2 | - | 9 | 1109 | |||
AF065922.2 | - | 8 | 982 | |||
AF065923.2 | - | 8 | 982 | |||
AY090567.1 | - | 8 | 932 | |||
BC062378.1 | - | 9 | 1121 | 4978648 | ||
M84373.1 | - | 9 | 1113 | |||
M87039.1 | - | 9 | 1184 | |||
M92649.1 | - | 9 | 1120 | |||
U43428.1 | - | 9 | 1000 |