Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013612.2 | NP_038640.2 | 3 | 338 | ||
GenBank mRNA | AK090303.1 | - | 3 | 339 | ||
AK144237.1 | - | 3 | 321 | |||
AK171343.1 | - | 3 | 267 | |||
AK171393.1 | - | 3 | 319 | |||
BC109137.1 | - | 3 | 252 | 40039390 | ||
BC109138.1 | - | 3 | 252 | 40039397 | ||
L13732.1 | - | 3 | 316 | |||
X75355.1 | - | 3 | 297 |