Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011019.3 | NP_035149.2 | 7 | 1027 | ||
GenBank mRNA | AB015978.1 | - | 7 | 1681 | ||
AF058805.1 | - | 7 | 997 | |||
AK087179.1 | - | 7 | 985 | |||
AK157508.1 | - | 2 | 197 | |||
BC137703.1 | - | 6 | 922 | 8860722 |