Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016776.2 | NP_058056.2 | 23 | 3100 | ||
GenBank mRNA | AK083908.1 | - | 23 | 3126 | ||
AK143469.1 | - | 23 | 3125 | |||
AK143487.1 | - | 23 | 3134 | |||
AK149747.1 | - | 18 | 2409 | |||
AK154016.1 | - | 21 | 3319 | |||
AK155074.1 | - | 23 | 3111 | |||
AK166533.1 | - | 12 | 1300 | |||
AK183304.1 | - | 3 | 184 | |||
BC004078.1 | - | 13 | 1584 | |||
BC015070.1 | - | 23 | 3101 | |||
BC048858.1 | - | 23 | 3100 | 5363740 | ||
BC052889.1 | - | 23 | 3121 | 30057169 | ||
BC060167.1 | - | 22 | 3196 | |||
U63648.1 | - | 23 | 3109 |