Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008832.2 | NP_032858.2 | 21 | 2639 | ||
NM_173021.3 | NP_766609.2 | 20 | 2462 | |||
GenBank mRNA | AK030426.1 | - | 21 | 2631 | ||
AK036440.1 | - | 20 | 2415 | |||
AK137497.1 | - | 20 | 2401 | |||
X74616.1 | - | 21 | 2535 |