Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011084.2 | NP_035214.2 | 2 | 272 | ||
NM_207683.2 | NP_997566.1 | 3018 | ||||
GenBank mRNA | AB008791.1 | - | 3018 | |||
AB008792.1 | - | 2 | 267 | |||
AK036916.1 | - | 2 | 83 | |||
AK054266.1 | - | 2 | 63 | |||
AK054271.1 | - | 2 | 148 | |||
AK054415.1 | - | 2 | 215 | |||
AK076317.1 | - | 2673 | ||||
BC061142.1 | - | 2 | 59 | |||
BC150813.1 | - | 3010 | 9087724 |