Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083110.2 | NP_001076579.1 | 2 | 1248 | ||
NM_001290555.1 | NP_001277484.1 | 2 | 1534 | |||
NM_001290556.1 | NP_001277485.1 | 3 | 708 | |||
NM_008853.3 | NP_032879.2 | 3 | 699 | |||
GenBank mRNA | AF335250.2 | - | 2 | 1217 | ||
AK005373.1 | - | 2 | 1155 | |||
AK135382.1 | - | 2 | 1233 | |||
AK145943.1 | - | 2 | 1491 | |||
BC008148.1 | - | 1 | 522 | |||
BC025975.1 | - | 2 | 1196 | 3983750 | ||
BC037616.1 | - | 2 | 1171 | 5401000 |