Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
AA591047, AU042537, EBS1, PCN, PLTN, Plec1
Chromosome Location:
Chr.15: 76170974 - 76231378 on Build GRCm38
UniGene ID:
Mm.234912
Species:
Mouse
siRNA ID
MSS207762
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163540.1
NP_001157012.1
2
768
NM_001163542.1
NP_001157014.1
2
249
NM_001163549.1
NP_001157021.1
2
331
NM_001164203.1
NP_001157675.1
2
236
NM_011117.2
NP_035247.2
2
249
NM_201385.2
NP_958787.2
2
184
NM_201386.2
NP_958788.2
2
245
NM_201387.3
NP_958789.3
2
274
NM_201388.2
NP_958790.2
2
236
NM_201389.2
NP_958791.2
2
768
NM_201390.2
NP_958792.2
2
183
NM_201391.2
NP_958793.2
2
331
NM_201392.2
NP_958794.2
2
157
NM_201393.2
NP_958795.2
2
274
NM_201394.2
NP_958796.2
2
331
GenBank mRNA
AF188006.1
-
356
AF188008.1
-
2
768
AF188009.1
-
2
768
AF188010.1
-
2
331
AF188011.1
-
2
331
AF188012.1
-
2
331
AF188013.1
-
2
249
AF188014.1
-
2
249
AF188015.1
-
2
183
AF188016.1
-
2
183
AF188017.1
-
2
271
AF188018.1
-
2
271
AF188019.1
-
2
184
AF188020.1
-
2
274
AF188021.1
-
2
157
AF188022.1
-
2
274
AF188023.1
-
2
236
AK017743.1
-
2
164
AK044559.1
-
293
AY480033.1
-
2
249
AY480034.1
-
2
184
AY480035.1
-
2
245
AY480036.1
-
2
274
AY480037.1
-
2
236
AY480038.1
-
2
768
AY480039.1
-
2
183
AY480040.1
-
2
331
AY480041.1
-
2
157
AY480042.1
-
2
274
AY480043.1
-
2
331
BC021810.1
-
2
318
BC099929.1
-
2
769
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to nutrient
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0