Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011132.2 | NP_035262.2 | 15 | 1715 | ||
GenBank mRNA | AF123502.1 | - | 15 | 1689 | ||
AK141674.1 | - | 15 | 1676 | |||
BC048166.1 | - | 15 | 1650 | 6410453 | ||
BC063246.1 | - | 15 | 1650 | 6410453 |