Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128605.1 | NP_001122077.1 | 7 | 932 | ||
NM_011183.3 | NP_035313.2 | 7 | 941 | |||
GenBank mRNA | AF038935.1 | - | 6 | 869 | ||
AK014706.1 | - | 6 | 876 | |||
AK148111.1 | - | 7 | 933 | |||
AK154119.1 | - | 7 | 877 | |||
AY864737.1 | - | 2 | 287 | |||
BC010403.1 | - | 6 | 887 | 4038761 | ||
BM935552.1 | - | 1 | 55 | |||
U57324.1 | - | 7 | 892 |