Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138668.2 | NP_619609.1 | 8 | 1002 | ||
GenBank mRNA | AK075795.1 | - | 8 | 997 | ||
AK078309.1 | - | 8 | 1003 | |||
AK160164.1 | - | 7 | 1077 | |||
BC005503.1 | - | 8 | 1003 | 3490744 |