Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138677.2 | NP_619618.1 | 5 | 1111 | ||
GenBank mRNA | AB042828.1 | - | 5 | 1100 | ||
AK030943.1 | - | 5 | 1112 | |||
AK037889.1 | - | 5 | 674 | |||
AK082920.1 | - | 5 | 1064 | |||
AK129090.1 | - | 5 | 1078 | |||
AK151668.1 | - | 5 | 1086 | |||
AK152724.1 | - | 5 | 1087 | |||
AK152979.1 | - | 5 | 1083 | |||
BC023237.1 | - | 5 | 1059 | 2654113 |