Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138950.2 | NP_620400.2 | 2 | 4014 | ||
GenBank mRNA | AK131146.1 | - | 2 | 614 | ||
AK143628.1 | - | 1 | 102 | |||
AK160585.1 | - | 1 | 56 | |||
AK172019.1 | - | 2 | 380 | |||
CN721357.1 | - | 2 | 524 |