Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008983.2 | NP_033009.1 | 14 | 2535 | ||
GenBank mRNA | AK052275.1 | - | 14 | 2489 | ||
AK083006.1 | - | 14 | 2499 | |||
BC138376.1 | - | 14 | 2501 | 8861396 | ||
BC145261.1 | - | 14 | 2501 |