Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166381.1 | NP_001159853.1 | 11 | 1321 | ||
NM_001166382.1 | NP_001159854.1 | 11 | 1318 | |||
NM_001166383.1 | NP_001159855.1 | 11 | 1321 | |||
NM_011236.2 | NP_035366.2 | 11 | 1321 | |||
GenBank mRNA | AK080702.1 | - | 11 | 1292 | ||
BC010673.1 | - | 10 | 1193 | |||
BC019123.1 | - | 11 | 1301 | 5003696 | ||
BE852444.1 | - | 2 | 374 | |||
U06837.1 | - | 11 | 1243 | |||
U12135.1 | - | 11 | 1287 | |||
Z32767.1 | - | 11 | 1315 |