Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011237.2 | NP_035367.1 | 5 | 507 | ||
GenBank mRNA | AF045663.1 | - | 5 | 507 | ||
AK134293.1 | - | 5 | 520 | |||
AK138973.1 | - | 5 | 512 | |||
AK165833.1 | - | 5 | 542 | |||
AK186361.1 | - | 3 | 233 | |||
AK219932.1 | - | 1 | 18 | |||
BC082556.1 | - | 5 | 446 | 30538342 |