Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011237.2 | NP_035367.1 | 3 | 306 | ||
GenBank mRNA | AF045663.1 | - | 3 | 306 | ||
AK134293.1 | - | 3 | 319 | |||
AK138973.1 | - | 3 | 311 | |||
AK165833.1 | - | 3 | 341 | |||
AK186361.1 | - | 1 | 32 | |||
BC082556.1 | - | 3 | 245 | 30538342 |