Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081052.2 | NP_001074521.1 | 6 | 2323 | ||
NM_001290526.1 | NP_001277455.1 | 7 | 2386 | |||
GenBank mRNA | BC147632.1 | - | 6 | 2323 | 9056622 | |
BC158016.1 | - | 7 | 2386 | 9007322 | ||
BC172134.1 | - | 7 | 2386 | 9054804 |