Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145834.1 | NP_001139306.1 | 16 | 2581 | ||
NM_001145835.1 | NP_001139307.1 | 16 | 2493 | |||
NM_001145836.1 | NP_001139308.1 | 16 | 2457 | |||
NM_009058.2 | NP_033084.2 | 16 | 2617 | |||
GenBank mRNA | AK129328.1 | - | 16 | 2458 | ||
AK141959.1 | - | 16 | 2448 | |||
AK149644.1 | - | 11 | 1352 | |||
AK159020.1 | - | 16 | 2450 | |||
AK159754.1 | - | 16 | 2463 | |||
AK188792.1 | - | 2 | 154 | |||
BC027225.1 | - | 16 | 2275 | 3486732 | ||
L07924.1 | - | 16 | 2453 |