Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 4 | 425 | ||
AK019171.1 | - | 4 | 440 | |||
AK090162.1 | - | 4 | 463 | |||
AK133924.1 | - | 4 | 525 | |||
AK145596.1 | - | 4 | 402 | |||
AK147057.1 | - | 4 | 385 | |||
AK151116.1 | - | 4 | 433 | |||
AK158987.1 | - | 4 | 442 | |||
AK159296.1 | - | 4 | 440 | |||
AK159778.1 | - | 4 | 434 | |||
AK159949.1 | - | 4 | 482 | |||
AK207872.1 | - | 4 | 363 | |||
BC003282.1 | - | 4 | 404 | 3582133 | ||
U95141.1 | - | 2 | 115 |