Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011280.2 | NP_035410.2 | 7 | 1543 | ||
GenBank mRNA | AF134811.1 | - | 7 | 1525 | ||
AF220121.1 | - | 7 | 1530 | |||
AI324236.1 | - | 1 | 703 | |||
AK011082.1 | - | 7 | 1543 | |||
BC051632.1 | - | 7 | 1516 | 6507331 |