Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176785.3 | NP_789742.2 | 1 | 1425 | ||
GenBank mRNA | AF536239.1 | - | 1 | 1370 | ||
AK030582.1 | - | 1 | 1396 | |||
AK040849.1 | - | 1 | 1372 | |||
AK159588.1 | - | 1 | 1391 | |||
AK172058.1 | - | 2 | 1176 | |||
AK206486.1 | - | 1 | 192 | |||
BC034055.1 | - | 1 | 1371 | 3673655 |