Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252313.1 | NP_001239242.1 | 36 | 5741 | ||
NM_011308.3 | NP_035438.3 | 36 | 5540 | |||
GenBank mRNA | AB093281.1 | - | 18 | 3481 | ||
AK136008.1 | - | 1 | 108 | |||
AK137632.1 | - | 10 | 1859 | |||
BC086657.1 | - | 36 | 5506 | 30545994 | ||
L78294.1 | - | 15 | 2548 | |||
U22016.1 | - | 2 | 295 | |||
U35312.1 | - | 35 | 5784 |