Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113326.1 | NP_001106797.1 | 5 | 749 | ||
NM_031195.2 | NP_112472.2 | 5 | 749 | |||
GenBank mRNA | AF203781.1 | - | 4 | 691 | ||
AK028480.1 | - | 5 | 743 | |||
AK089178.1 | - | 5 | 743 | |||
AK150518.1 | - | 5 | 750 | |||
AK151403.1 | - | 5 | 723 | |||
AK151855.1 | - | 5 | 750 | |||
AK154806.1 | - | 5 | 726 | |||
AK155362.1 | - | 5 | 745 | |||
AK155589.1 | - | 5 | 743 | |||
BC003814.1 | - | 5 | 720 | 3594142 | ||
D13382.1 | - | 4 | 691 | |||
L04274.1 | - | 5 | 696 | |||
L04275.1 | - | 5 | 696 |