Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011340.3 | NP_035470.3 | 8 | 1290 | ||
GenBank mRNA | AA111321.1 | - | 1 | 94 | ||
AF017057.1 | - | 8 | 1188 | |||
AF036164.1 | - | 8 | 1188 | |||
BC019852.1 | - | 8 | 1200 | 5123884 | ||
D87975.1 | - | 8 | 1187 |