Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198968.2 | NP_001185897.1 | 38 | 4950 | ||
NM_011365.4 | NP_035495.2 | 37 | 4869 | |||
GenBank mRNA | AF132479.1 | - | 36 | 4612 | ||
AK122480.1 | - | 32 | 4256 | |||
BC037119.1 | - | 1 | 495 | |||
BC138262.1 | - | 38 | 4927 | 8861281 | ||
BC171950.1 | - | 38 | 4927 | 9054236 | ||
BE650375.1 | - | 4 | 370 |