Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011369.2 | NP_035499.1 | 6 | 813 | ||
GenBank mRNA | AF017152.1 | - | 6 | 813 | ||
AK137496.1 | - | 6 | 829 | |||
AK144966.1 | - | 6 | 836 | |||
AK149580.1 | - | 6 | 830 | |||
AK162943.1 | - | 6 | 827 | |||
BC070455.1 | - | 6 | 833 | 30606785 |