Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011382.2 | NP_035512.1 | 2 | 1163 | ||
GenBank mRNA | BC137931.1 | - | 2 | 1146 | 8860950 | |
BC137934.1 | - | 2 | 1146 | 8860953 | ||
D50416.1 | - | 2 | 1163 | |||
D50417.1 | - | 4 | 1078 | |||
D50418.1 | - | 4 | 936 |