Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015748.3 | NP_056563.2 | 30, 31 | 3658 | ||
GenBank mRNA | AF144627.1 | - | 30, 31 | 3683 | ||
AK122376.1 | - | 30, 31 | 3424 | |||
BC057131.1 | - | 30, 31 | 3464 | 6827560 | ||
BC062091.1 | - | 30, 31 | 3464 | 6827560 |