Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 17 | 1766 | ||
NM_011431.3 | NP_035561.1 | 17 | 1763 | |||
GenBank mRNA | AK052242.1 | - | 17 | 1721 | ||
AK150158.1 | - | 17 | 1734 | |||
AK153473.1 | - | 17 | 1735 | |||
AK154878.1 | - | 17 | 1745 | |||
AK165624.1 | - | 17 | 1737 | |||
AK172875.1 | - | 17 | 1717 | |||
AK178609.1 | - | 2 | 97 | |||
BC012636.1 | - | 4 | 478 | |||
BC052674.1 | - | 17 | 1739 | 30061580 | ||
BC054778.1 | - | 17 | 1729 | 6810127 | ||
U97079.1 | - | 17 | 1705 |