Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146010.1 | NP_001139482.1 | 9, 10 | 1075 | ||
NM_199012.2 | NP_950177.2 | 9, 10 | 1075 | |||
GenBank mRNA | AK122368.1 | - | 9, 10 | 967 | ||
AK140322.1 | - | 9, 10 | 1054 | |||
BC060647.1 | - | 9, 10 | 1037 | 6854041 |