Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011461.3 | NP_035591.3 | 4, 5 | 363 | ||
GenBank mRNA | AF098863.2 | - | 4, 5 | 362 | ||
AK137522.1 | - | 4, 5 | 366 | |||
AK156024.1 | - | 4, 5 | 364 | |||
AK156892.1 | - | 4, 5 | 364 | |||
AK157188.1 | - | 4, 5 | 365 | |||
BC064038.1 | - | 3, 4 | 324 | 3824951 |