Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039388.2 | NP_001034477.1 | 2 | 355 | ||
NM_001039389.2 | NP_001034478.1 | 2 | 276 | |||
NM_172445.3 | NP_766033.1 | 2 | 276 | |||
GenBank mRNA | AK014128.1 | - | 2 | 252 | ||
AK032582.1 | - | 2 | 238 | |||
AK036203.1 | - | 2 | 272 | |||
AK044740.1 | - | 2 | 256 | |||
AK129254.1 | - | 2 | 232 | |||
AK146209.1 | - | 2 | 251 | |||
BB611667.1 | - | 2 | 355 | |||
BC046236.1 | - | 1 | 103 | 6817089 | ||
BC048819.1 | - | 2 | 200 | |||
CJ134699.1 | - | 2 | 277 |